Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esyt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011843.2 | NP_035973.1 | 18 | 2069 | ||
GenBank mRNA | AF098634.1 | - | 9 | 862 | ||
AK044656.1 | - | 18 | 2079 | |||
AK054147.1 | - | 9 | 853 | |||
AK080094.1 | - | 18 | 2082 | |||
AK088421.1 | - | 18 | 2083 | |||
AK142877.1 | - | 1 | 1558 | |||
BC011482.1 | - | 18 | 2069 | 4165946 |