Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oas1g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF480417.1 | - | 3 | 652 | ||
BC018470.1 | - | 636 | 3493001 | |||
BC043339.1 | - | 3 | 667 | 5004267 | ||
X58077.1 | - | 579 |
Gene Symbol: Oas1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145211.2 | NP_660212.2 | 3 | 665 | ||
GenBank mRNA | AK149613.1 | - | 3 | 626 | ||
BC013715.1 | - | 3 | 647 | 3257027 | ||
BC057878.1 | - | 3 | 624 | 5348858 | ||
M33863.1 | - | 651 | ||||
X04958.1 | - | 3 | 651 |