Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oas1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083925.1 | NP_001077394.1 | 6 | 1124 | ||
GenBank mRNA | AB067529.1 | - | 6 | 1124 | ||
AF328926.1 | - | 6 | 1124 | |||
AF418004.1 | - | 6 | 1124 | |||
AF418005.1 | - | 6 | 1124 | |||
AF418006.1 | - | 6 | 1124 | |||
AF418007.1 | - | 6 | 1124 | |||
AF418008.1 | - | 6 | 1124 | |||
AF418009.1 | - | 6 | 1124 | |||
AF466822.1 | - | 6 | 1081 | |||
AF466823.1 | - | 6 | 1081 | |||
AK080152.1 | - | 6 | 1117 | |||
BC012877.1 | - | 6 | 1116 | 4016177 |