Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Papss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289477.1 | NP_001276406.1 | 12 | 1815 | ||
NM_001289478.1 | NP_001276407.1 | 12 | 2002 | |||
NM_001289479.1 | NP_001276408.1 | 13 | 2113 | |||
NM_011863.2 | NP_035993.1 | 11 | 1825 | |||
GenBank mRNA | AK146507.1 | - | 11 | 1698 | ||
AK153296.1 | - | 11 | 1695 | |||
AK160875.1 | - | 13 | 2017 | |||
AK168982.1 | - | 11 | 1698 | |||
AK169117.1 | - | 12 | 1877 | |||
AK169124.1 | - | 11 | 1692 | |||
AK169204.1 | - | 12 | 1877 | |||
AK210041.1 | - | 1 | 65 | |||
BC066055.1 | - | 12 | 1881 | 6833756 | ||
U34883.1 | - | 11 | 1702 |