Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Papss2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201470.1 | NP_001188399.1 | 9 | 1234 | ||
NM_011864.3 | NP_035994.2 | 10 | 1270 | |||
GenBank mRNA | AF052453.1 | - | 10 | 1251 | ||
AF085144.1 | - | 9 | 1166 | |||
AK128962.1 | - | 9 | 1255 | |||
AK137675.1 | - | 9 | 1160 | |||
BC090997.1 | - | 10 | 1270 | 30121331 |