Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pebp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018858.2 | NP_061346.2 | 3 | 302 | ||
GenBank mRNA | AB046417.1 | - | 3 | 304 | ||
AF300422.1 | - | 3 | 281 | |||
AK008037.1 | - | 318 | ||||
AK088212.1 | - | 3 | 307 | |||
AK143897.1 | - | 3 | 334 | |||
AK149815.1 | - | 3 | 318 | |||
AK159842.1 | - | 3 | 307 | |||
AK168790.1 | - | 3 | 303 | |||
AK179859.1 | - | 1 | 29 | |||
AK182328.1 | - | 3 | 192 | |||
BC008169.1 | - | 3 | 336 | 3257403 | ||
BC083063.1 | - | 3 | 302 | 5362125 | ||
BC089332.1 | - | 3 | 285 | 30359654 | ||
U43206.1 | - | 3 | 300 |