Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290707.1 | NP_001277636.1 | 14 | 1649 | ||
NM_011866.2 | NP_035996.2 | 13 | 1603 | |||
GenBank mRNA | AF110507.1 | - | 13 | 1474 | ||
AK039249.1 | - | 14 | 1649 | |||
AK147690.1 | - | 12 | 1520 | |||
AK162221.1 | - | 13 | 1258 | |||
AK162804.1 | - | 11 | 1144 | |||
AK166310.1 | - | 5 | 615 | |||
AY360383.1 | - | 13 | 1603 | |||
BC113201.1 | - | 13 | 1469 | 40048238 |