Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cib1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291275.1 | NP_001278204.1 | 4 | 471 | ||
NM_001291276.1 | NP_001278205.1 | 2 | 342 | |||
NM_011870.5 | NP_036000.1 | 4 | 486 | |||
GenBank mRNA | AB006463.1 | - | 4 | 267 | ||
AF173010.1 | - | 4 | 265 | |||
AK010345.1 | - | 4 | 283 | |||
AK027915.1 | - | 4 | 298 | |||
AK088450.1 | - | 2 | 157 | |||
AK152805.1 | - | 4 | 318 | |||
AK165472.1 | - | 4 | 271 | |||
BB843350.1 | - | 2 | 121 | |||
BC003714.1 | - | 4 | 272 | 3599986 | ||
BC054385.1 | - | 4 | 291 | 6476137 | ||
CF104297.1 | - | 4 | 494 |