Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rai2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103367.1 | NP_001096837.1 | 2 | 389 | ||
NM_198409.3 | NP_940801.2 | 2 | 467 | |||
GenBank mRNA | AK143442.1 | - | 2 | 357 | ||
BB857753.1 | - | 2 | 390 | |||
BC061184.1 | - | 2 | 393 | 30300326 | ||
X76652.1 | - | 1 | 133 |