Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011879.2 | NP_036009.1 | 15 | 1448 | ||
GenBank mRNA | AJ006130.1 | - | 15 | 1331 | ||
AK082356.1 | - | 15 | 1380 | |||
AK087976.1 | - | 15 | 1394 | |||
BC006706.1 | - | 13 | 1806 | |||
BC014739.1 | - | 15 | 1379 | 4920184 | ||
BC026569.1 | - | 15 | 1379 | 4982612 | ||
BC029148.1 | - | 13 | 2342 | |||
BC029671.1 | - | 15 | 1391 | 5364953 | ||
BC029694.1 | - | 15 | 1388 | 5369477 | ||
BC033356.1 | - | 15 | 1375 | 5366230 |