Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199003.1 | NP_001185932.1 | 10, 11 | 1019 | ||
NM_011880.3 | NP_036010.2 | 10, 11 | 1019 | |||
GenBank mRNA | AF011360.1 | - | 10, 11 | 864 | ||
BC051133.1 | - | 10, 11 | 974 | 6588901 |