Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnasel | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011882.2 | NP_036012.1 | 4 | 1857 | ||
GenBank mRNA | AF281045.1 | - | 2 | 1401 | ||
AK053125.1 | - | 3 | 1556 | |||
AK054205.1 | - | 2 | 2352 | |||
AK140815.1 | - | 4 | 1858 | |||
BC003873.1 | - | 4 | 1666 | |||
BC138013.1 | - | 1 | 1274 | 8861032 |