Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abce1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015751.2 | NP_056566.2 | 5 | 551 | ||
GenBank mRNA | AK049299.1 | - | 5 | 552 | ||
AK133748.1 | - | 5 | 482 | |||
AK134738.1 | - | 5 | 484 | |||
AK144711.1 | - | 5 | 480 | |||
AK147151.1 | - | 5 | 489 | |||
AK167130.1 | - | 5 | 499 | |||
AK167370.1 | - | 5 | 489 | |||
AK167704.1 | - | 5 | 450 | |||
BC005422.1 | - | 5 | 472 | 3489529 | ||
BC066794.1 | - | 4 | 254 | |||
BC066836.1 | - | 4 | 254 | |||
U90446.1 | - | 5 | 440 |