Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204269.1 | NP_001191198.1 | 3 | 948 | ||
NM_004974.3 | NP_004965.1 | 3 | 994 | |||
XM_011541396.2 | XP_011539698.1 | 3 | 988 | |||
XM_011541397.2 | XP_011539699.1 | 4 | 1269 | |||
XM_011541398.2 | XP_011539700.1 | 3 | 1036 | |||
XM_011541399.2 | XP_011539701.1 | 3 | 1077 | |||
XM_011541400.2 | XP_011539702.1 | 3 | 1007 | |||
XM_017001213.1 | XP_016856702.1 | 3 | 969 | |||
GenBank mRNA | AK290061.1 | - | 3 | 994 | ||
BC043564.1 | - | 3 | 948 | 5212949 | ||
L02752.1 | - | 1 | 373 |