Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rngtt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF025653.1 | - | 5 | 432 | ||
AF034568.1 | - | 5 | 486 | |||
AK002922.1 | - | 5 | 535 | |||
AK079488.1 | - | 5 | 532 | |||
AK142016.1 | - | 5 | 529 | |||
AK150497.1 | - | 5 | 539 | |||
AK151463.1 | - | 5 | 553 | |||
AK160007.1 | - | 5 | 541 | |||
AK160706.1 | - | 5 | 519 | |||
AK169305.1 | - | 5 | 505 |