Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scamp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011886.2 | NP_036016.2 | 5 | 603 | ||
GenBank mRNA | AF005036.1 | - | 5 | 510 | ||
AF295403.1 | - | 5 | 508 | |||
AK135335.1 | - | 5 | 587 | |||
AK146204.1 | - | 5 | 605 | |||
AK160549.1 | - | 5 | 588 | |||
BC002021.1 | - | 5 | 601 | 3489293 |