Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011889.2 | NP_036019.2 | 6, 7 | 780 | ||
GenBank mRNA | AK030668.1 | - | 6, 7 | 748 | ||
AK046722.1 | - | 6, 7 | 873 | |||
AK141915.1 | - | 6, 7 | 707 | |||
AK164861.1 | - | 6, 7 | 877 | |||
AK169916.1 | - | 6, 7 | 901 | |||
AK220520.1 | - | 4, 5 | 579 | |||
BC055738.1 | - | 6, 7 | 709 | 6413697 |