Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sigirr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023059.3 | NP_075546.2 | 7 | 1239 | ||
GenBank mRNA | AF113795.1 | - | 6 | 756 | ||
AF239957.1 | - | 6 | 750 | |||
AK133518.1 | - | 7 | 864 | |||
AK145889.1 | - | 7 | 1242 | |||
BC010806.1 | - | 6 | 761 | 4240425 | ||
BC094069.1 | - | 7 | 845 | 4974962 |