Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sigirr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023059.3 | NP_075546.2 | 5, 6 | 1082 | ||
GenBank mRNA | AF113795.1 | - | 4, 5 | 599 | ||
AF239957.1 | - | 4, 5 | 593 | |||
AK133518.1 | - | 5, 6 | 707 | |||
AK145889.1 | - | 5, 6 | 1085 | |||
BC010806.1 | - | 4, 5 | 604 | 4240425 | ||
BC094069.1 | - | 5, 6 | 688 | 4974962 |