Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sufu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025391.2 | NP_001020562.1 | 10 | 1494 | ||
NM_015752.3 | NP_056567.2 | 10 | 1497 | |||
GenBank mRNA | AF134893.1 | - | 10 | 1437 | ||
AJ131692.1 | - | 10 | 1495 | |||
AJ308626.1 | - | 10 | 1428 | |||
AK016976.1 | - | 10 | 1475 | |||
AK156410.1 | - | 10 | 1459 | |||
AK215049.1 | - | 1 | 68 | |||
BC048168.1 | - | 10 | 1467 | 6821477 | ||
BC056997.1 | - | 10 | 1467 | 6821477 |