Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011906.2 | NP_036036.2 | 10 | 1099 | ||
GenBank mRNA | AF051098.1 | - | 10 | 906 | ||
AK089317.1 | - | 10 | 970 | |||
AK155116.1 | - | 10 | 1043 | |||
AK164913.1 | - | 10 | 856 | |||
AK172045.1 | - | 10 | 1030 | |||
BC010244.1 | - | 8 | 655 | |||
BC028977.1 | - | 10 | 1013 | 5363085 | ||
BC057296.1 | - | 10 | 1032 |