Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbck1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083921.1 | NP_001077390.1 | 4, 5 | 823 | ||
NM_019705.3 | NP_062679.2 | 5, 6 | 664 | |||
GenBank mRNA | AF124663.1 | - | 3, 4 | 433 | ||
AK085559.1 | - | 4, 5 | 812 | |||
AK088591.1 | - | 4, 5 | 785 | |||
AK146176.1 | - | 5, 6 | 838 | |||
AK166075.1 | - | 4, 5 | 791 | |||
AK209721.1 | - | 1, 2 | 47 | |||
BC034555.1 | - | 5, 6 | 660 | 4192310 |