Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbck1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083921.1 | NP_001077390.1 | 2 | 503 | ||
NM_019705.3 | NP_062679.2 | 3 | 344 | |||
GenBank mRNA | AF124663.1 | - | 1 | 113 | ||
AK085559.1 | - | 2 | 492 | |||
AK088591.1 | - | 2 | 465 | |||
AK146176.1 | - | 2 | 462 | |||
AK166075.1 | - | 2 | 471 | |||
AK181405.1 | - | 1 | 71 | |||
AK182273.1 | - | 1 | 71 | |||
AK210389.1 | - | 1 | 71 | |||
AK213806.1 | - | 1 | 71 | |||
BC034555.1 | - | 3 | 340 | 4192310 | ||
BY267597.1 | - | 3 | 323 | |||
CX238505.1 | - | 2 | 503 |