Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023137.3 | NP_075626.1 | 2 | 449 | ||
GenBank mRNA | AK008116.1 | - | 2 | 450 | ||
AK008552.1 | - | 2 | 422 | |||
BC027627.2 | - | 2 | 422 | 1068927 | ||
BC036383.1 | - | 2 | 422 | 1347593 | ||
CT010257.1 | - | 2 | 389 |