Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xrn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011917.2 | NP_036047.2 | 17 | 1622 | ||
GenBank mRNA | AK004800.1 | - | 17 | 1597 | ||
AK011711.1 | - | 17 | 1620 | |||
AK031247.1 | - | 17 | 1665 | |||
AK053643.1 | - | 17 | 1615 | |||
AK167019.1 | - | 17 | 1642 | |||
AK168037.1 | - | 17 | 1668 | |||
BC004028.1 | - | 9 | 793 | 3495063 | ||
BC052395.1 | - | 6 | 538 | |||
BC052823.1 | - | 17 | 1483 | |||
BC054743.1 | - | 17 | 1622 | 6400974 | ||
D38517.1 | - | 17 | 1560 |