Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ldb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039071.2 | NP_001034160.1 | 6 | 945 | ||
NM_001039072.2 | NP_001034161.1 | 7 | 813 | |||
NM_001039073.2 | NP_001034162.1 | 8 | 828 | |||
NM_001039074.2 | NP_001034163.1 | 7 | 859 | |||
NM_001039075.2 | NP_001034164.1 | 8 | 828 | |||
NM_001039076.2 | NP_001034165.1 | 6 | 945 | |||
NM_011918.4 | NP_036048.3 | 6 | 945 | |||
GenBank mRNA | AF114378.2 | - | 6 | 911 | ||
AF114379.2 | - | 8 | 792 | |||
AF228057.1 | - | 5 | 838 | |||
AF228058.1 | - | 5 | 838 | |||
AJ005621.1 | - | 8 | 821 | |||
AK004020.1 | - | 8 | 836 | |||
AK084461.1 | - | 6 | 925 | |||
AK137181.1 | - | 6 | 928 | |||
AK142292.1 | - | 6 | 897 | |||
AK172980.1 | - | 7 | 859 | |||
AY206011.1 | - | 5 | 862 | |||
AY206012.1 | - | 6 | 911 | |||
AY206013.1 | - | 7 | 777 | |||
AY206015.1 | - | 7 | 744 | |||
BC099596.1 | - | 8 | 795 | |||
BC138793.1 | - | 5 | 879 | 8861815 | ||
BC145420.1 | - | 5 | 879 | 9053978 |