Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136089.2 | NP_001129561.1 | 10 | 918 | ||
NM_011922.3 | NP_036052.2 | 9 | 858 | |||
GenBank mRNA | AJ238978.1 | - | 10 | 818 | ||
AK008772.1 | - | 10 | 841 | |||
AK008808.1 | - | 10 | 842 | |||
AK020288.1 | - | 9 | 783 | |||
AK131880.1 | - | 10 | 838 | |||
AK168715.1 | - | 10 | 822 | |||
BC125319.1 | - | 10 | 790 | 40129834 | ||
BC132207.1 | - | 10 | 790 | 40130484 |