Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnpda1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011937.2 | NP_036067.2 | 2 | 190 | ||
GenBank mRNA | AF088903.1 | - | 2 | 148 | ||
AF160355.1 | - | 2 | 136 | |||
AK080610.1 | - | 2 | 138 | |||
AK084247.1 | - | 1 | 820 | |||
AK166069.1 | - | 2 | 140 | |||
AK172879.1 | - | 1 | 53 | |||
AK200561.1 | - | 1 | 50 | |||
BC014800.1 | - | 2 | 121 | 4220980 | ||
BY092839.1 | - | 2 | 165 | |||
CJ048539.1 | - | 2 | 191 |