Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grk6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038018.4 | NP_001033107.1 | 14 | 1669 | ||
NM_001112711.2 | NP_001106182.1 | 14 | 1669 | |||
NM_001286063.1 | NP_001272992.1 | 14 | 1649 | |||
NM_001286064.1 | NP_001272993.1 | 14 | 1654 | |||
NM_001286065.1 | NP_001272994.1 | 14 | 1567 | |||
NM_001286066.1 | NP_001272995.1 | 14 | 1662 | |||
NM_011938.4 | NP_036068.2 | 14 | 1669 | |||
GenBank mRNA | AF040747.1 | - | 14 | 1539 | ||
AF040748.1 | - | 14 | 1539 | |||
AF040749.1 | - | 14 | 1539 | |||
AK051405.1 | - | 14 | 1651 | |||
AK146119.1 | - | 14 | 1442 | |||
AK161142.1 | - | 12 | 1301 | |||
BC057206.1 | - | 14 | 1643 | |||
BC075719.1 | - | 14 | 1550 | 5031455 | ||
CA528447.1 | - | 3 | 268 | |||
Y15797.1 | - | 14 | 1491 | |||
Y15798.1 | - | 14 | 1513 | |||
Y15799.1 | - | 14 | 1496 | |||
Y15800.1 | - | 14 | 1484 |