Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map4k1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008279.2 | NP_032305.2 | 24 | 1880 | ||
GenBank mRNA | AK143215.1 | - | 25 | 1964 | ||
AK146273.1 | - | 24 | 1864 | |||
BC005433.1 | - | 24 | 1873 | 3494470 | ||
BC024832.1 | - | 11 | 737 | |||
Y09010.1 | - | 24 | 1916 |