Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUSB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK096764.1 | - | 4 | 2341 | ||
AK300223.1 | - | 12 | 1872 | |||
AK303819.1 | - | 10 | 1542 | |||
CD014092.1 | - | 11 | 1780 | |||
CD014093.1 | - | 10 | 1638 | |||
CD014094.1 | - | 11 | 1880 | |||
M15182.1 | - | 12 | 1978 |