Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004977.2 | NP_004968.2 | 2 | 1907 | ||
XM_006723203.2 | XP_006723266.1 | 2 | 1612 | |||
XM_011526925.2 | XP_011525227.1 | 2 | 1612 | |||
XM_011526926.1 | XP_011525228.1 | 2 | 1612 | |||
XM_011526928.2 | XP_011525230.1 | 2 | 1612 | |||
GenBank mRNA | AB208930.1 | - | 1 | 713 | ||
AF055989.1 | - | 2 | 1907 |