Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163671.1 | NP_001157143.1 | 8 | 1011 | ||
NM_001163672.1 | NP_001157144.1 | 8 | 1011 | |||
NM_016961.3 | NP_058657.1 | 8 | 1011 | |||
NM_207692.2 | NP_997575.2 | 8 | 1011 | |||
GenBank mRNA | AB005664.1 | - | 7 | 785 | ||
AF052466.1 | - | 7 | 747 | |||
AF052467.1 | - | 7 | 747 | |||
AF052468.1 | - | 7 | 747 | |||
AF052469.1 | - | 7 | 747 | |||
AK031959.1 | - | 8 | 1010 | |||
AK031974.1 | - | 6 | 565 | |||
AK145083.1 | - | 8 | 986 | |||
BC028341.1 | - | 8 | 993 | 4192796 |