Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nubp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011955.2 | NP_036085.1 | 11 | 956 | ||
GenBank mRNA | AF114170.1 | - | 11 | 933 | ||
AK020141.1 | - | 6 | 493 | |||
AK157212.1 | - | 11 | 948 | |||
BC031430.1 | - | 11 | 947 | |||
BC047926.1 | - | 9 | 2131 | |||
BC055436.1 | - | 11 | 944 | 3660324 | ||
BU614991.1 | - | 4 | 237 | |||
CX210766.1 | - | 8 | 644 |