Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plod3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011962.3 | NP_036092.1 | 10 | 1698 | ||
GenBank mRNA | AF046783.1 | - | 10 | 1106 | ||
AK013195.1 | - | 10 | 1244 | |||
AK033360.1 | - | 10 | 1243 | |||
AK053320.1 | - | 10 | 1340 | |||
AK088948.1 | - | 10 | 1115 | |||
AK157741.1 | - | 9 | 1348 | |||
AK183180.1 | - | 2 | 49 | |||
AK183516.1 | - | 2 | 49 | |||
BC043047.1 | - | 10 | 1697 | 5703967 | ||
BC054734.1 | - | 10 | 1698 | 5720686 |