Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psma1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011965.2 | NP_036095.1 | 3 | 219 | ||
GenBank mRNA | AF060088.1 | - | 3 | 143 | ||
AJ272019.1 | - | 3 | 172 | |||
AK135326.1 | - | 3 | 157 | |||
AK137054.1 | - | 3 | 161 | |||
AK144784.1 | - | 3 | 174 | |||
AK161611.1 | - | 3 | 161 | |||
AK162277.1 | - | 3 | 166 | |||
AK167689.1 | - | 3 | 157 | |||
BC005762.1 | - | 3 | 156 | 2655483 |