Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUSB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000181.3 | NP_000172.2 | 4 | 729 | ||
NM_001293104.1 | NP_001280033.1 | 3 | 544 | |||
NR_120531.1 | 4 | 729 | ||||
XM_005250297.3 | XP_005250354.1 | 4 | 724 | |||
XM_011516113.1 | XP_011514415.1 | 3 | 460 | |||
XM_011516114.1 | XP_011514416.1 | 3 | 388 | |||
XM_017012091.1 | XP_016867580.1 | 3 | 460 | |||
XM_017012092.1 | XP_016867581.1 | 3 | 460 | |||
XM_017012093.1 | XP_016867582.1 | 3 | 388 | |||
GenBank mRNA | AK223406.1 | - | 4 | 628 | ||
AK311273.1 | - | 3 | 443 | |||
BC014142.2 | - | 4 | 599 | 4662011 | ||
CD014092.1 | - | 3 | 426 | |||
CD014094.1 | - | 4 | 611 | |||
M15182.1 | - | 4 | 624 |
Gene Symbol: GUSBP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024448.2 | 3 | 596 | |||
GenBank mRNA | AK056875.1 | - | 3 | 529 | ||
AK091837.1 | - | 3 | 454 | |||
AK300413.1 | - | 3 | 498 | |||
BX538181.1 | - | 3 | 444 |