Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mok | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011973.2 | NP_036103.1 | 4 | 431 | ||
GenBank mRNA | AB022695.1 | - | 4 | 431 |