Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc27a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009512.2 | NP_033538.2 | 6 | 1530 | ||
GenBank mRNA | AF072760.1 | - | 6 | 1319 | ||
AJ223959.1 | - | 6 | 1488 | |||
BC013272.1 | - | 6 | 1495 | 4193807 | ||
BC013335.1 | - | 6 | 1495 | 4193567 | ||
BC115421.2 | - | 6 | 1470 | 40093476 | ||
BC145823.1 | - | 6 | 1470 | 40131126 | ||
BC145825.1 | - | 6 | 1470 | 40131128 |