Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Homer3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146153.1 | NP_001139625.1 | 3 | 278 | ||
NM_011984.2 | NP_036114.2 | 3 | 278 | |||
GenBank mRNA | AF093261.1 | - | 1 | 105 | ||
AK155463.1 | - | 1 | 2457 | |||
BC005773.1 | - | 3 | 227 | 3602414 | ||
BC095961.1 | - | 3 | 233 | |||
CX208892.1 | - | 3 | 257 |