Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ror2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013846.3 | NP_038874.3 | 9 | 1882 | ||
GenBank mRNA | AB010384.1 | - | 9 | 1650 | ||
AK031112.1 | - | 9 | 1852 | |||
AK031789.1 | - | 9 | 1639 | |||
AK083538.1 | - | 9 | 1858 | |||
AK084752.1 | - | 9 | 1848 | |||
AK157705.1 | - | 9 | 1898 | |||
BC030848.1 | - | 7 | 1223 | |||
BC070436.1 | - | 9 | 1882 | 30535615 |