Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ror2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013846.3 | NP_038874.3 | 4, 5 | 726 | ||
GenBank mRNA | AB010384.1 | - | 4, 5 | 494 | ||
AK031112.1 | - | 4, 5 | 696 | |||
AK031789.1 | - | 4, 5 | 484 | |||
AK083538.1 | - | 4, 5 | 702 | |||
AK084752.1 | - | 4, 5 | 692 | |||
AK157705.1 | - | 4, 5 | 742 | |||
BC030848.1 | - | 2, 3 | 67 | |||
BC070436.1 | - | 4, 5 | 726 | 30535615 |