Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ror2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013846.3 | NP_038874.3 | 9 | 2167 | ||
GenBank mRNA | AB010384.1 | - | 9 | 1935 | ||
AK031112.1 | - | 9 | 2137 | |||
AK031789.1 | - | 9 | 1924 | |||
AK083538.1 | - | 9 | 2143 | |||
AK084752.1 | - | 9 | 2133 | |||
AK157705.1 | - | 9 | 2183 | |||
BC030848.1 | - | 7 | 1508 | |||
BC070436.1 | - | 9 | 2167 | 30535615 |