Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla2g10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291009.1 | NP_001277938.1 | 3 | 864 | ||
NM_011987.3 | NP_036117.1 | 3 | 864 | |||
GenBank mRNA | AF166097.2 | - | 3 | 466 | ||
AF210429.1 | - | 3 | 292 | |||
AK076921.1 | - | 3 | 264 | |||
BC028879.1 | - | 3 | 324 | 4218273 | ||
KC789644.1 | - | 3 | 310 |