Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc27a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011989.4 | NP_036119.1 | 3 | 529 | ||
GenBank mRNA | AJ251113.1 | - | 2 | 242 | ||
AK036919.1 | - | 3 | 531 | |||
AK155313.1 | - | 3 | 499 | |||
AK155388.1 | - | 3 | 499 | |||
AK170377.1 | - | 3 | 533 | |||
BC023114.1 | - | 3 | 380 | 4158956 | ||
CT010312.1 | - | 2 | 242 |