Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc27a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011989.4 | NP_036119.1 | 7 | 1254 | ||
GenBank mRNA | AF072759.1 | - | 5 | 556 | ||
AJ251113.1 | - | 6 | 967 | |||
AK009292.1 | - | 1 | 41 | |||
AK033519.1 | - | 3 | 198 | |||
AK036919.1 | - | 7 | 1256 | |||
AK155313.1 | - | 7 | 1223 | |||
AK155388.1 | - | 7 | 1224 | |||
AK170377.1 | - | 7 | 1258 | |||
AK213719.1 | - | 1 | 37 | |||
BC023114.1 | - | 7 | 1105 | 4158956 | ||
CT010312.1 | - | 6 | 967 |