Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011990.2 | NP_036120.1 | 2 | 652 | ||
GenBank mRNA | AB022345.1 | - | 2 | 619 | ||
AK037490.1 | - | 2 | 653 | |||
AK037742.1 | - | 2 | 627 | |||
AK076322.1 | - | 2 | 627 | |||
AK089202.1 | - | 2 | 625 | |||
AK162233.1 | - | 2 | 425 | |||
AK163168.1 | - | 2 | 626 | |||
AK165599.1 | - | 2 | 628 | |||
AK171192.1 | - | 2 | 650 | |||
AK171252.1 | - | 2 | 626 | |||
AY766236.1 | - | 2 | 619 | |||
AY766237.1 | - | 619 | ||||
BB630675.1 | - | 2 | 653 | |||
BC141402.1 | - | 2 | 549 | 9056015 | ||
BC141408.1 | - | 2 | 549 | 9056021 |