Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rcn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011992.2 | NP_036122.2 | 6 | 904 | ||
GenBank mRNA | AF049125.1 | - | 6 | 893 | ||
AK041996.1 | - | 901 | ||||
AK076153.1 | - | 6 | 731 | |||
AK077486.1 | - | 6 | 880 | |||
AK169765.1 | - | 6 | 878 | |||
BC132320.1 | - | 6 | 869 | 40130597 | ||
BC145668.1 | - | 6 | 869 | 40130971 |