Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpysl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011993.4 | NP_036123.3 | 12 | 1458 | ||
GenBank mRNA | AB006715.1 | - | 12 | 1387 | ||
AK090197.1 | - | 11 | 1986 | |||
AK145638.1 | - | 11 | 1561 | |||
AK165694.1 | - | 12 | 1459 | |||
BC046333.1 | - | 11 | 3011 | |||
BC090674.1 | - | 1430 | ||||
BC138573.1 | - | 12 | 1376 | 8861593 | ||
BC138574.1 | - | 12 | 1376 | 8861594 | ||
Y09079.1 | - | 12 | 1437 |