Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pclo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110796.2 | NP_001104266.1 | 5 | 6328 | ||
NM_011995.4 | NP_036125.4 | 5 | 6328 | |||
GenBank mRNA | AB083477.1 | - | 5 | 6253 | ||
AB083478.1 | - | 5 | 6253 | |||
Y19185.2 | - | 6 | 6238 | |||
Y19186.2 | - | 6 | 6238 |